Platinium(IV) oxide, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Platinum dioxide, Adam's type catalyst
M-Clarity™ quality level = MQ200
459925-250MG 459925-1G 459925-5G
SIAL459925-250MGEA 91.7 EUR
SIAL459925-250MG SIAL459925-1G SIAL459925-5G
Platinium(IV) oxide, Sigma-Aldrich®
Platinium (IV) oxide
Formula: O₂Pt
MW: 227.08 g/mol
Melting Pt: 450 °C
Density: 11.8 g/cm³ (20 °C)
MDL Number: MFCD00011184
CAS Number: 1314-15-4
EINECS: 215-223-0
UN: 1479
ADR: 5.1,II
Merck Index: 12,07686