Iron(II) sulphide 99.9% (trace metals basis) -100 mesh
Supplier: Thermo Scientific
Synonyms: Ferrous sulphide
315870100 315870500
ACRO315870100EA 82.2 EUR
ACRO315870100 ACRO315870500
Iron(II) sulphide 99.9% (trace metals basis) -100 mesh
Iron (II) sulphide
Formula: FeS
MW: 87.91 g/mol
Melting Pt: 1195 °C
Density: 4.84 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011013
CAS Number: 1317-37-9
EINECS: 215-268-6
ADR: 9,III
Merck Index: 12,04106
Order Now
Specification Test Results
Appearance Grey to brown to black Powder
Total trace metal impurities ≤0.1 %