Cobalt(II) sulphate heptahydrate ≥98%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Cobalt sulphate heptahydrate, Cobaltous sulphate heptahydrate
A16201.0E A16201.22 A16201.36
A16201.0EEA 497 EUR
A16201.0E A16201.22 A16201.36
Cobalt(II) sulphate heptahydrate ≥98%
Cobalt (II) sulphate heptahydrate
Purity: 98% CAS No.: 10026-24-1 Molecular Formula: CoO4S.7H2O Formula Weight: 281.10 (154.99anhy) Odour: Odourless pH: 4 Merck Reference: 14,2448 UN No.: 3077 MDL No.: MFCD00149658
Formula: CoSO₄·7H₂O
MW: 281.1 g/mol
Melting Pt: 735
Density: 2.03
Storage Temperature: Ambient
MDL Number: MFCD00149658
CAS Number: 10026-24-1
EINECS: 233-334-2
UN: 3077
ADR: 9 III
Merck Index: 12,02510