Nickel sulphate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Nickel (II) sulphate
M-Clarity™ quality level = MQ100
656895-10G 656895-50G
SIAL656895-10GEA 90.1 EUR
SIAL656895-10G SIAL656895-50G
Nickel sulphate, Sigma-Aldrich®
Nickel sulphate
Formula: NiO₄S
MW: 154.75 g/mol
Melting Pt: 848
Density: 3.543 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011146
CAS Number: 7786-81-4
EINECS: 232-104-9
UN: 3077
ADR: 9 III
Merck Index: 12,06600