Succinic anhydride
Supplier: MP Biomedicals
Warning
Synonyms: Dihydrofuran-2,5-dione
02102970.5 0210297001
ICNA02102970.5EA 138 EUR
ICNA02102970.5 ICNA0210297001
Succinic anhydride
Succinic anhydride
Formula: C₄H₄O₃
MW: 100.07 g/mol
Boiling Pt: 261
Melting Pt: 120
Density: 1.503 g/cm³ (20 °C)
Flash Pt: 157
MDL Number: MFCD00005525
CAS Number: 108-30-5
EINECS: 203-570-0
UN: 3261
ADR: 8 III
Merck Index: 12,09039