Succinic anhydride, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Dihydrofuran-2,5-dione
M-Clarity™ quality level = MQ200
239690-250G 239690-50G
SIAL239690-250GEA 83.2 EUR
SIAL239690-250G SIAL239690-50G
Succinic anhydride, Sigma-Aldrich®
Succinic anhydride
Formula: C₄H₄O₃
Boiling Pt: 261 °C (1013 hPa)
Melting Pt: 120 °C
Density: 1.503 g/cm³ (20 °C)
Flash Pt: 157 °C
MDL Number: MFCD00005525
CAS Number: 108-30-5
EINECS: 203-570-0
UN: 3261
ADR: 8,III
Merck Index: 12,09039