Span® 60, Sigma-Aldrich®

Supplier: Merck

Synonyms: Arlacel 60, Sorbitan monooctadecanoate, Sorbitan stearate, (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol monooctadecanoate, Sorbitan monostearate

M-Clarity™ quality level = MQ200

S7010-1KG S7010-250G
SIALS7010-1KGEA 159 EUR
SIALS7010-1KG SIALS7010-250G
Span® 60, Sigma-Aldrich®
Span® 60
Formula: C₂₄H₄₆O₆
MW: 430.63 g/mol
Boiling Pt: ∼100 °C (1013 hPa)
Melting Pt: 53 °C
Density: 1 g/cm³ (20 °C)
Flash Pt: >149 °C
Storage Temperature: Ambient
MDL Number: MFCD00005366
CAS Number: 1338-41-6
EINECS: 215-664-9
Merck Index: 12,08872

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR