Dehydroacetic acid, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 3-Acetyl-6-methyl-2H-pyran-2,4(3H)-dione, 3-Acetyl-4-hydroxy-6-methyl-2H-pyran-2-one

30740-100G 30740-500G
SIAL30740-100GEA 43.8 EUR
SIAL30740-100G SIAL30740-500G
Dehydroacetic acid, Sigma-Aldrich®
Dehydracetic acid
Formula: C₈H₈O₄
MW: 168.15 g/mol
Boiling Pt: 333 °C (1013 hPa)
Melting Pt: 108 °C
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00066709
CAS Number: 520-45-6
EINECS: 208-293-9
Merck Index: 13,02885

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR