Pentacene, Sigma-Aldrich®
Supplier: Merck
Synonyms:
2,3,6,7-Dibenzanthracene
MQ100
P1802-5G
P1802-100MG
P1802-1G
SIALP1802-5GEA
1360
EUR
SIALP1802-5G
SIALP1802-100MG
SIALP1802-1G
Pentacene, Sigma-Aldrich®
Pentacene
Formula:
C₂₂H₁₄ MW: 278.35 g/mol Melting Pt: 300 Density: 1.3 g/cm³ (20 °C) |
MDL Number:
MFCD00003710 CAS Number: 135-48-8 EINECS: 205-193-7 Merck Index: 12,07240 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...