Perylene, Sigma-Aldrich®
Supplier: Merck
394475-1G 394475-5G
SIAL394475-1GEA 118 EUR
SIAL394475-1G SIAL394475-5G
Perylene, Sigma-Aldrich®
Perylene
Formula: C₂₀H₁₂
MW: 252.32 g/mol
Boiling Pt: 443
Melting Pt: 273…278 °C
Density: 1.362 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004142
CAS Number: 198-55-0
EINECS: 205-900-9
Merck Index: 12,07325