Phenanthrene, Sigma-Aldrich®
Supplier: Merck
Warning
MQ100
P11409-500G P11409-100G P11409-25G
SIALP11409-500GEA 685 EUR
SIALP11409-500G SIALP11409-100G SIALP11409-25G
Phenanthrene, Sigma-Aldrich®
Phenanthrene
Formula: C₁₄H₁₀
MW: 178.23 g/mol
Boiling Pt: 340
Melting Pt: 97…99 °C
Density: 1.19 g/cm³ (20 °C)
Flash Pt: 153
Storage Temperature: Ambient
MDL Number: MFCD00001168
CAS Number: 85-01-8
EINECS: 201-581-5
UN: 3077
ADR: 9 III
Merck Index: 12,07354
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92527d856e1fe8582e54bf3812e1580ee0b91b05&vid=d8ec397dac001e49a7958f12aacb577cd11954da&prid=web&clid=SAPDAM