3,8-Diamino-6-phenylphenanthridine 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
B24049.03
B24049.06
B24049.14
B24049.03EA
77.9
EUR
B24049.03
B24049.06
B24049.14
3,8-Diamino-6-phenylphenanthridine 98%
3,8-Diamino-6-phenylphenanthridine
Purity: 98%
CAS No.: 52009-64-0
Molecular Formula: C19H15N3
Formula Weight: 285.35
Melting Point: 196-198°
MDL No.: MFCD00075152
Formula:
C₁₉H₁₅N₃ Melting Pt: 196…198 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075152 CAS Number: 52009-64-0 EINECS: 257-602-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...