18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane), Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

28125-100G 28125-25G 28125-5G
SIAL28125-100GEA 272 EUR
SIAL28125-100G SIAL28125-25G SIAL28125-5G
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane), Sigma-Aldrich®
18-Crown-6 (1,4,7,10,13,16-Hexaoxacyclooctadecane)
Formula: C₁₂H₂₄O₆
MW: 264.32 g/mol
Boiling Pt: 396 °C (1013 hPa)
Melting Pt: 37…39 °C
Density: 1.237 g/cm³ (25 °C)
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00005113
CAS Number: 17455-13-9
EINECS: 241-473-5
Merck Index: 13,02630

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR