15-Crown-5 (1,4,7,10,13-pentaoxacyclopentadecane, lead ionophore V), Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 15-Crown-5, 1,4,7,10,13-Pentaoxacyclopentadecane
M-Clarity™ quality level = MQ200
188832-25G 188832-1G 188832-5G
SIAL188832-25GEA 161 EUR
SIAL188832-25G SIAL188832-1G SIAL188832-5G
15-Crown-5 (1,4,7,10,13-pentaoxacyclopentadecane, lead ionophore V), Sigma-Aldrich®
15-Crown-5 (1,4,7,10,13-Pentaoxacyclopentadecane, Lead ionophore V)
Formula: C₁₀H₂₀O₅
MW: 220.27 g/mol
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: –20 °C
Density: 1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005110
CAS Number: 33100-27-5
EINECS: 251-379-6