2,6-Dichloropurine 97%
Supplier: Thermo Scientific
Warning
Synonyms: 2,6-Dichloro-9H-purine
113700010 113700050
ACRO113700010EA 30 EUR
ACRO113700010 ACRO113700050
2,6-Dichloropurine 97%
2,6-Dichloropurine
Formula: C₅H₂Cl₂N₄
MW: 189 g/mol
Melting Pt: 184…186 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00077725
CAS Number: 5451-40-1
EINECS: 226-681-6
Order Now
Specification Test Results
Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
GC ≥97.5 %