Theobromine, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
3,7-Dimethyl-3,7-dihydro-1H-purine-2,6-dione, 3,7-Dimethylxanthine, 3,7-Dihydro-3,7-dimethyl-1H-purine-2,6-dione, 2,6-Dihydroxy-3,7-dimethylpurine
M-Clarity™ quality level = MQ200
T4500-25G
T4500-100G
SIALT4500-25GEA
48.7
EUR
SIALT4500-25G
SIALT4500-100G
Theobromine, Sigma-Aldrich®
Theobromine
Formula:
C₇H₈N₄O₂ MW: 180.17 g/mol Melting Pt: 350 °C Density: 1.419 g/cm³ (25 °C) |
MDL Number:
MFCD00022830 CAS Number: 83-67-0 EINECS: 201-494-2 Merck Index: 12,09418 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...