6-Benzylaminopurine, Sigma-Aldrich®

Supplier: Merck

Synonyms: 6-BAP, 6-Benzyladenine, N6-Benzyladenine, Benzyl(purin-6-yl)amine

13151-1G-F 13151-25G-F 13151-5G-F
SIAL13151-1G-FEA 31.9 EUR
SIAL13151-1G-F SIAL13151-25G-F SIAL13151-5G-F
6-Benzylaminopurine, Sigma-Aldrich®
6-Benzylaminopurine
Formula: C₁₂H₁₁N₅
MW: 225.25 g/mol
Melting Pt: 230…234 °C
MDL Number: MFCD00005572
CAS Number: 1214-39-7
EINECS: 214-927-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR