2-Chloropyrazine, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Chloropyrazine
M-Clarity™ quality level = MQ100
132489-25G
132489-5G
SIAL132489-25GEA
39.9
EUR
SIAL132489-25G
SIAL132489-5G
2-Chloropyrazine, Sigma-Aldrich®
2-Chloropyrazine
Formula:
C₄H₃ClN₂ MW: 114.53 g/mol Boiling Pt: 153 °C (1013 hPa) Density: 1.28 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006124 CAS Number: 14508-49-7 EINECS: 238-517-0 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...