Pyrazine, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
W401501-100G-K W401501-1KG-K
SIALW401501-100G-KEA 81.8 EUR
SIALW401501-100G-K SIALW401501-1KG-K
Pyrazine, Sigma-Aldrich®
Pyrazine
Formula: C₄H₄N₂
Boiling Pt: 115 °C (1013 hPa)
Melting Pt: 52 °C
Density: 1.0311 g/cm³ (60.9 °C)
MDL Number: MFCD00006122
CAS Number: 290-37-9
EINECS: 206-027-6
UN: 1325
ADR: 4.1,III
Merck Index: 13,08047