2,3,5-Trimethylpyrazine, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
W324426-100G-K W324426-1KG-K
SIALW324426-100G-KEA 515 EUR
SIALW324426-100G-K SIALW324426-1KG-K
2,3,5-Trimethylpyrazine, Sigma-Aldrich®
2,3,5-Trimethylpyrazine
Formula: C₇H₁₀N₂
Boiling Pt: 171 °C (1013 hPa)
Density: 0.9958 g/cm³ (0 °C)
MDL Number: MFCD00006145
CAS Number: 14667-55-1
EINECS: 238-712-0
UN: 1993
ADR: 3,III