2,3-Diethyl-5-methylpyrazine, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ300
W333603-100G-K W333603-1KG-K
SIALW333603-100G-KEA 178 EUR
SIALW333603-100G-K SIALW333603-1KG-K
2,3-Diethyl-5-methylpyrazine, Sigma-Aldrich®
2,3-Diethyl-5-methylpyrazine
Formula: C₉H₁₄N₂
MW: 150.22 g/mol
Boiling Pt: 95
Density: 0.95 g/cm³ (20 °C)
MDL Number: MFCD00009635
CAS Number: 18138-04-0
EINECS: 242-024-6