2,3,5,6-Tetramethylpyrazine, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ300
W323705-1KG-K
W323705-250G-K
W323705-5KG-K
SIALW323705-1KG-KEA
399
EUR
SIALW323705-1KG-K
SIALW323705-250G-K
SIALW323705-5KG-K
2,3,5,6-Tetramethylpyrazine, Sigma-Aldrich®
2,3,5,6-Tetramethylpyrazine
Formula:
C₈H₁₂N₂ MW: 136.2 g/mol Boiling Pt: 190 °C (1013 hPa) Density: 1.15 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006146 CAS Number: 1124-11-4 EINECS: 214-391-2 |