1H-Pyrazole, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Pyrazole, 1H-Pyrazole
M-Clarity™ quality level = MQ200
P56607-25G P56607-100G P56607-5G
SIALP56607-25GEA 56 EUR
SIALP56607-25G SIALP56607-100G SIALP56607-5G
1H-Pyrazole, Sigma-Aldrich®
1H-Pyrazole
Formula: C₃H₄N₂
MW: 68.08 g/mol
Boiling Pt: 186 °C (1013 hPa)
Melting Pt: 66…68 °C
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005234
CAS Number: 288-13-1
EINECS: 206-017-1
Merck Index: 12,08144