Pyrene, Sigma-Aldrich®
Supplier: Merck
82648-10G 82648-1G
SIAL82648-10GEA 188 EUR
SIAL82648-10G SIAL82648-1G
Pyrene, Sigma-Aldrich®
Pyrene
Formula: C₁₆H₁₀
MW: 202.26 g/mol
Boiling Pt: 404
Melting Pt: 148…150 °C
Density: 1.27 g/cm³ (20 °C)
Flash Pt: 200…220 °C
MDL Number: MFCD00004136
CAS Number: 129-00-0
EINECS: 204-927-3
Merck Index: 12,08147
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b03eb6b87b60315a282988993f066fd2d3af129&vid=d3f1ffa103d650d2e9f49139db04842dfa6ca0ff&prid=web&clid=SAPDAM