3,6-Dichloropyridazine
Supplier: Thermo Scientific
Warning
BTB05810.1 BTB05810.10 BTB05810.50
MAYBBTB05810.1EA 0 EUR
MAYBBTB05810.1 MAYBBTB05810.10 MAYBBTB05810.50
3,6-Dichloropyridazine
3,6-Dichloropyridazine
Formula: C₄H₂Cl₂N₂
MW: 148.98 g/mol
Boiling Pt: 89…91 °C (0.2 mmHg)
Melting Pt: 70 °C
Density: 1.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006466
CAS Number: 141-30-0
EINECS: 205-478-6
UN: 2811
ADR: 6.1,III