2-Bromopyridine, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

B80100-100G B80100-25G B80100-500G
SIALB80100-100GEA 95.9 EUR
SIALB80100-100G SIALB80100-25G SIALB80100-500G
2-Bromopyridine, Sigma-Aldrich®
2-Bromopyridine
Formula: C₅H₄BrN
MW: 158 g/mol
Boiling Pt: 192 °C (1013 hPa)
Melting Pt: –40.1 °C
Density: 1.62 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006219
CAS Number: 109-04-6
EINECS: 203-641-6
UN: 2810
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR