3-Bromopyridine, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ200

B80208-100G B80208-25G B80208-5G
SIALB80208-100GEA 254 EUR
SIALB80208-100G SIALB80208-25G SIALB80208-5G
3-Bromopyridine, Sigma-Aldrich®
3-Bromopyridine
Formula: C₅H₄BrN
Boiling Pt: 173 °C (1013 hPa)
Melting Pt: –27 °C
Density: 1.62 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006373
CAS Number: 626-55-1
EINECS: 210-952-0
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR