5-Bromo-2-chloropyridine, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

551902-25G 551902-5G
SIAL551902-25GEA 118 EUR
SIAL551902-25G SIAL551902-5G
5-Bromo-2-chloropyridine, Sigma-Aldrich®
5-Bromo-2-chloropyridine
Formula: C₅H₃BrClN
MW: 192.44 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 71 °C
MDL Number: MFCD01318951
CAS Number: 53939-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR