5-Bromonicotinic acid
Supplier: Thermo Scientific
Synonyms:
3-Bromo-5-pyridine carboxylic acid, 5-Bromopyridine-3-carboxylic acid, 5-Bromo-3-pyridinecarboxylic acid
SB01732.1
SB01732.10
SB01732.50
MAYBSB01732.1EA
0
EUR
MAYBSB01732.1
MAYBSB01732.10
MAYBSB01732.50
5-Bromonicotinic acid
5-Bromonicotinic acid
Formula:
C₆H₄BrNO₂ MW: 202.01 g/mol Boiling Pt: 300 Melting Pt: 184…185 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009783 CAS Number: 20826-04-4 EINECS: 244-065-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...