5-Bromonicotinic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: 3-Bromo-5-pyridine carboxylic acid, 5-Bromopyridine-3-carboxylic acid, 5-Bromo-3-pyridinecarboxylic acid

M-Clarity™ quality level = MQ100

228435-10G 228435-50G
SIAL228435-10GEA 70 EUR
SIAL228435-10G SIAL228435-50G
5-Bromonicotinic acid, Sigma-Aldrich®
5-Bromonicotinic acid
Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
Boiling Pt: 300
Melting Pt: 184…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00009783
CAS Number: 20826-04-4
EINECS: 244-065-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR