2-Bromo-5-iodopyridine, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

652598-25G 652598-5G
SIAL652598-25GEA 131 EUR
SIAL652598-25G SIAL652598-5G
2-Bromo-5-iodopyridine, Sigma-Aldrich®
2-Bromo-5-iodopyridine
Formula: C₅H₃BrIN
MW: 283.89 g/mol
MDL Number: MFCD03095201
CAS Number: 73290-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR