2-Amino-5-bromopyridine, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 5-Bromo-2-pyridylamine, 5-Bromopyridin-2-amine
M-Clarity™ quality level = MQ100
122858-25G 122858-5G
SIAL122858-25GEA 73.1 EUR
SIAL122858-25G SIAL122858-5G
2-Amino-5-bromopyridine, Sigma-Aldrich®
2-Amino-5-bromopyridine
Formula: C₅H₅BrN₂
MW: 173.01 g/mol
Boiling Pt: 244 °C (1013 hPa)
Melting Pt: 136…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00006323
CAS Number: 1072-97-5
EINECS: 214-019-9