4-Amino-3-bromopyridine, Sigma-Aldrich®

Supplier: Merck

Synonyms: 3-Bromo-4-pyridylamine, 3-Bromopyridin-4-amine

685224-1G 685224-5G
SIAL685224-1GEA 73.8 EUR
SIAL685224-1G SIAL685224-5G
4-Amino-3-bromopyridine, Sigma-Aldrich®
4-Amino-3-bromopyridine
Formula: C₅H₅BrN₂
MW: 173.01 g/mol
Boiling Pt: 244 °C (1013 hPa)
Melting Pt: 181…182.5 °C
MDL Number: MFCD02068297
CAS Number: 13534-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR