2-Bromo-3-hydroxypyridine, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-Bromo-3-pyridinol
M-Clarity™ quality level = MQ100
116173-25G 116173-5G
SIAL116173-25GEA 149 EUR
SIAL116173-25G SIAL116173-5G
2-Bromo-3-hydroxypyridine, Sigma-Aldrich®
2-Bromo-3-hydroxypyridine
Formula: C₅H₄BrNO
Boiling Pt: 232 °C (1013 hPa)
Melting Pt: 185…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00006220
CAS Number: 6602-32-0
EINECS: 229-547-5