2,3,5-Trichloropyridine, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

SIAL384275-50GEA 83.6 EUR
SIAL384275-50G
2,3,5-Trichloropyridine, Sigma-Aldrich®
2,3,5-Trichloropyridine
Formula: C₅H₂Cl₃N
MW: 182,44 g/mol
MDL Number: MFCD00043007
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR