2-Chloropyridine, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
C69802-100G
C69802-500G
C69802-5G
SIALC69802-100GEA
50.6
EUR
SIALC69802-100G
SIALC69802-500G
SIALC69802-5G
2-Chloropyridine, Sigma-Aldrich®
2-Chloropyridine
Formula:
C₅H₄ClN MW: 113.55 g/mol Boiling Pt: 166 °C (1013 hPa) Melting Pt: –46 °C Density: 1.205 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006228 CAS Number: 109-09-1 EINECS: 203-646-3 UN: 2822 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...