3-Chloropyridine, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
C70002-25G C70002-50G
SIALC70002-25GEA 57.5 EUR
SIALC70002-25G SIALC70002-50G
3-Chloropyridine, Sigma-Aldrich®
3-Chloropyridine
Formula: C₅H₄ClN
MW: 113.55 g/mol
Boiling Pt: 148 °C (1013 hPa)
Melting Pt: –0.5 °C
Density: 1.194 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006375
CAS Number: 626-60-8
EINECS: 210-955-7
UN: 1100
ADR: 3,I