2-Chloronicotinonitrile ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
A14818.06
A14818.14
A14818.22
A14818.06EA
33.8
EUR
A14818.06
A14818.14
A14818.22
2-Chloronicotinonitrile ≥98%
2-Chloronicotinonitrile
Purity: 98%
CAS No.: 6602-54-6
Molecular Formula: C6H3ClN2
Formula Weight: 138.56
Melting Point: 105-109°
Boiling Point: 112°/1mm
UN No.: 3439
MDL No.: MFCD00014628
Formula:
C₆H₃ClN₂ Boiling Pt: 112 °C (1 mmHg) Melting Pt: 105…109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014628 CAS Number: 6602-54-6 EINECS: 229-550-1 UN: 3439 ADR: 6.1,III |