6-Chloronicotinamide

Supplier: Thermo Scientific

Synonyms: 6-Chloropyridine-3-carboxamide

RF04494.1 RF04494.10 RF04494.50
MAYBRF04494.1EA 0 EUR
MAYBRF04494.1 MAYBRF04494.10 MAYBRF04494.50
6-Chloronicotinamide
6-Chloronicotinamide
Formula: C₆H₅ClN₂O
MW: 156.57 g/mol
MDL Number: MFCD00006242
CAS Number: 6271-78-9
EINECS: 228-456-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR