6-Chloronicotinic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 6-Chloropyridine-3-carboxylic acid, 6-Chloro-3-pyridinecarboxylic acid
M-Clarity™ quality level = MQ200
156353-100G 156353-25G 156353-5G
SIAL156353-100GEA 62.6 EUR
SIAL156353-100G SIAL156353-25G SIAL156353-5G
6-Chloronicotinic acid, Sigma-Aldrich®
6-Chloronicotinic acid
Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
Boiling Pt: 284 °C (1013 hPa)
Melting Pt: 188…189 °C
Storage Temperature: Ambient
MDL Number: MFCD00006241
CAS Number: 5326-23-8
EINECS: 226-201-5