2-Chloro-3-fluoropyridine, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

636576-25G 636576-5G
SIAL636576-25GEA 227 EUR
SIAL636576-25G SIAL636576-5G
2-Chloro-3-fluoropyridine, Sigma-Aldrich®
2-Chloro-3-fluoropyridine
Formula: C₅H₃ClFN
MW: 131.54 g/mol
MDL Number: MFCD03095302
CAS Number: 17282-04-1
UN: 2810
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR