2-Amino-5-chloropyridine 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 5-Chloropyridin-2-amine, 5-Chloro-2-pyridylamine
A12063.14 A12063.22 A12063.36
A12063.14EA 47.8 EUR
A12063.14 A12063.22 A12063.36
2-Amino-5-chloropyridine 98%
2-Amino-5-chloropyridine
Purity: 98% CAS No.: 1072-98-6 Molecular Formula: C5H5ClN2 Formula Weight: 128.56 Flash Point: 160°(320°F) Melting Point: 135-138° Boiling Point: 127-129°/21mm Beilstein Reference: 108735 MDL No.: MFCD00006324
Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Boiling Pt: 127…129 °C (21 mmHg)
Melting Pt: 135…138 °C
Density: 1.514 g/cm³ (20 °C)
Flash Pt: 160 °C (320 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006324
CAS Number: 1072-98-6
EINECS: 214-020-4