2-Amino-5-chloropyridine, Sigma-Aldrich®
Supplier: Merck
Synonyms: 5-Chloropyridin-2-amine, 5-Chloro-2-pyridylamine
M-Clarity™ quality level = MQ100
A46803-100G A46803-25G
SIALA46803-100GEA 79.2 EUR
SIALA46803-100G SIALA46803-25G
2-Amino-5-chloropyridine, Sigma-Aldrich®
2-Amino-5-chloropyridine
Formula: C₅H₅ClN₂
Boiling Pt: 241 °C (1013 hPa)
Melting Pt: 129…131 °C
Density: 1.514 g/cm³ (20 °C)
MDL Number: MFCD00006324
CAS Number: 1072-98-6
EINECS: 214-020-4