3-Amino-2-chloropyridine, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-Chloro-3-pyridylamine, 2-Chloropyridin-3-amine
M-Clarity™ quality level = MQ100
A46900-10G A46900-50G
SIALA46900-10GEA 52.2 EUR
SIALA46900-10G SIALA46900-50G
3-Amino-2-chloropyridine, Sigma-Aldrich®
3-Amino-2-chloropyridine
Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Boiling Pt: 263 °C (1013 hPa)
Melting Pt: 78…79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006238
CAS Number: 6298-19-7
EINECS: 228-572-9