5-Amino-2-chloropyridine, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-Chloro-5-pyridylamine, 6-Chloropyridin-3-amine, 2-Chloropyridin-5-amine
M-Clarity™ quality level = MQ200
188778-1G 188778-5G
SIAL188778-1GEA 26.1 EUR
SIAL188778-1G SIAL188778-5G
5-Amino-2-chloropyridine, Sigma-Aldrich®
5-Amino-2-chloropyridine
Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Boiling Pt: 225 °C (1013 hPa)
Melting Pt: 83 °C
MDL Number: MFCD00006243
CAS Number: 5350-93-6
EINECS: 226-322-3