2-Chloro-6-methoxypyridine, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
152730-25G 152730-5G
SIAL152730-25GEA 125 EUR
SIAL152730-25G SIAL152730-5G
2-Chloro-6-methoxypyridine, Sigma-Aldrich®
2-Chloro-6-methoxypyridine
Formula: C₆H₆ClNO
MW: 143.57 g/mol
Boiling Pt: 39…40 °C (0.3 mmHg)
Density: 1.207 g/cm³ (20 °C)
MDL Number: MFCD00006265
CAS Number: 17228-64-7
EINECS: 241-264-9