Nicotinonitrile 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 3-Pyridinecarbonitrile, 3-Cyanopyridine, Pyridine-3-carbonitrile
110860050 110861000 110865000 110860025
ACRO110860050EA 23.7 EUR
ACRO110860050 ACRO110861000 ACRO110865000 ACRO110860025
Nicotinonitrile 98%
Nicotinonitrile
Formula: C₆H₄N₂
MW: 104.11 g/mol
Boiling Pt: 206.2
Melting Pt: 47…51 °C
Density: 1.159 g/cm³
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00087130
CAS Number: 100-54-9
EINECS: 202-863-0
Order Now
Specification Test Results
Appearance Colorless to white to yellow Crystals or crystalline mass and/or chunks
Infrared spectrum Conforms
Melting point 47 to 51 °C
Water ≤0.2 %
GC ≥97.5 %