Picolinic acid
Supplier: Thermo Scientific
Warning
CC04001DA CC04001EA
MAYBCC04001DAEA 0 EUR
MAYBCC04001DA MAYBCC04001EA
Picolinic acid
Picolinic acid
Formula: C₆H₅NO₂
Boiling Pt: 293 °C (1013 hPa)
Melting Pt: 134…137 °C
Density: 1.49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006293
CAS Number: 98-98-6
EINECS: 202-719-7
Merck Index: 12,07557