Picolinic acid
Supplier: Merck
Warning
Synonyms: Pyridine-2-carboxylic acid, 2-Picolinic acid, 2-Pyridinecarboxylic acid
M-Clarity™ quality level = MQ200
P42800-100G P42800-500G P42800-5G P42800-5KG P42800-2KG
SIALP42800-100GEA 74.1 EUR
SIALP42800-100G SIALP42800-500G SIALP42800-5G SIALP42800-5KG SIALP42800-2KG
Picolinic acid
Picolinic acid
Formula: C₆H₅NO₂
MW: 123.11 g/mol
Boiling Pt: 293 °C (1013 hPa)
Melting Pt: 134…137 °C
Density: 1.49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006293
CAS Number: 98-98-6
EINECS: 202-719-7
Merck Index: 12,07557