Nicotinic acid (Niacin)
Supplier: Thermo Scientific
Warning
Synonyms: 3-Pyridinecarboxylic acid, Pyridine-3-carboxylic acid, 3-Picolinic acid, Niacin, Nicotinic acid, Vitamin B3
CC04101.1 CC04101.10
MAYBCC04101.1EA 0 EUR
MAYBCC04101.1 MAYBCC04101.10
Nicotinic acid (Niacin)
Nicotinic acid (Niacin)
Formula: C₆H₅NO₂
MW: 123.11 g/mol
Boiling Pt: 258
Melting Pt: 142
Density: 1.473 g/cm³ (20 °C)
Flash Pt: 193
Storage Temperature: Ambient
MDL Number: MFCD00006391
CAS Number: 59-67-6
EINECS: 200-441-0
Merck Index: 13,06552